Learn More
Isonicotinic acid, 99%
CAS: 55-22-1 | C6H5NO2 | 123.111 g/mol
$51.90 - $570.19
Chemical Identifiers
| CAS | 55-22-1 |
|---|---|
| Molecular Formula | C6H5NO2 |
| Molecular Weight (g/mol) | 123.111 |
| MDL Number | MFCD00006429 |
| InChI Key | TWBYWOBDOCUKOW-UHFFFAOYSA-N |
| Synonym | isonicotinic acid, 4-pyridinecarboxylic acid, 4-picolinic acid, 4-carboxypyridine, p-pyridinecarboxylic acid, gamma-picolinic acid, gamma-pyridinecarboxylic acid, acide iso-nicotinique, 1,4-dihydroisonicotinic acid, acide iso-nicotinique french |
| PubChem CID | 5922 |
| ChEBI | CHEBI:6032 |
| IUPAC Name | pyridine-4-carboxylic acid |
| SMILES | C1=CN=CC=C1C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1810922
|
Thermo Scientific Chemicals
A1810922 |
100 g |
N/A
|
|
|||||
|
AAA1810936
|
Thermo Scientific Chemicals
A1810936 |
500 g |
N/A
|
|
|||||
|
AAA181090E
|
Thermo Scientific Chemicals
A181090E |
2500 g |
N/A
|
|
|||||
Description
It acts as an useful isomer of nicotinic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt acts as an useful isomer of nicotinic acid.
Solubility
Soluble in water (6 mg/ml), and 2 N NH4OH (5%). Insoluble in ether, benzene, and boiling alcohol.
Notes
Store away from oxidizing agents and reducing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
| 55-22-1 | |
| 123.111 | |
| TWBYWOBDOCUKOW-UHFFFAOYSA-N | |
| 5922 | |
| pyridine-4-carboxylic acid |
| C6H5NO2 | |
| MFCD00006429 | |
| isonicotinic acid, 4-pyridinecarboxylic acid, 4-picolinic acid, 4-carboxypyridine, p-pyridinecarboxylic acid, gamma-picolinic acid, gamma-pyridinecarboxylic acid, acide iso-nicotinique, 1,4-dihydroisonicotinic acid, acide iso-nicotinique french | |
| CHEBI:6032 | |
| C1=CN=CC=C1C(=O)O |
Specifications
| 55-22-1 | |
| 3.0 to 4.0 | |
| C6H5NO2 | |
| 100 g | |
| 14,5187 | |
| Soluble in water (6mg/ml),and 2 N NH4OH (5%). Insoluble in ether,benzene,and boiling alcohol. | |
| C1=CN=CC=C1C(=O)O | |
| 123.111 | |
| CHEBI:6032 | |
| 99% |
| ∼310°C (sublimation) | |
| 260°C (15 mmHg) | |
| MFCD00006429 | |
| 109599 | |
| isonicotinic acid, 4-pyridinecarboxylic acid, 4-picolinic acid, 4-carboxypyridine, p-pyridinecarboxylic acid, gamma-picolinic acid, gamma-pyridinecarboxylic acid, acide iso-nicotinique, 1,4-dihydroisonicotinic acid, acide iso-nicotinique french | |
| TWBYWOBDOCUKOW-UHFFFAOYSA-N | |
| pyridine-4-carboxylic acid | |
| 5922 | |
| 123.11 | |
| Isonicotinic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 200-228-2
RTECSNumber : NS1103000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only