missing translation for 'onlineSavingsMsg'
Learn More

Isonicotinic acid, 99%

CAS: 55-22-1 | C6H5NO2 | 123.111 g/mol

Supplier:  Thermo Scientific Chemicals A181090E

 View more versions of this product

Catalog No. AAA181090E


Only null left
Explore available promotions

Description

Description

It acts as an useful isomer of nicotinic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It acts as an useful isomer of nicotinic acid.

Solubility
Soluble in water (6 mg/ml), and 2 N NH4OH (5%). Insoluble in ether, benzene, and boiling alcohol.

Notes
Store away from oxidizing agents and reducing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

Isonicotinic acid
∼310°C (sublimation)
260°C (15 mmHg)
MFCD00006429
109599
isonicotinic acid, 4-pyridinecarboxylic acid, 4-picolinic acid, 4-carboxypyridine, p-pyridinecarboxylic acid, gamma-picolinic acid, gamma-pyridinecarboxylic acid, acide iso-nicotinique, 1,4-dihydroisonicotinic acid, acide iso-nicotinique french
TWBYWOBDOCUKOW-UHFFFAOYSA-N
pyridine-4-carboxylic acid
5922
123.11
55-22-1
3.0 to 4.0
C6H5NO2
2500 g
14,5187
Soluble in water (6mg/ml),and 2 N NH4OH (5%). Insoluble in ether,benzene,and boiling alcohol.
C1=CN=CC=C1C(=O)O
123.111
CHEBI:6032
99%

Chemical Identifiers

55-22-1
123.111
TWBYWOBDOCUKOW-UHFFFAOYSA-N
5922
pyridine-4-carboxylic acid
C6H5NO2
MFCD00006429
isonicotinic acid, 4-pyridinecarboxylic acid, 4-picolinic acid, 4-carboxypyridine, p-pyridinecarboxylic acid, gamma-picolinic acid, gamma-pyridinecarboxylic acid, acide iso-nicotinique, 1,4-dihydroisonicotinic acid, acide iso-nicotinique french
CHEBI:6032
C1=CN=CC=C1C(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 200-228-2

RTECSNumber : NS1103000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only