missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Isomannide | |
| 86°C | |
| C6H10O4 | |
| 25 g | |
| KLDXJTOLSGUMSJ-UHFFFAOYNA-N | |
| hexahydrofuro[3,2-b]furan-3,6-diol | |
| 12304540 | |
| ≥98.0% (GC) |
| 641-74-7 | |
| 150°C | |
| MFCD10698761,MFCD32202826,MFCD10698761 | |
| 1,4:3,6-Dianhydromannitol | |
| OC1COC2C(O)COC12 | |
| 146.14 | |
| 146.14 | |
| Crystalline Powder |
Chemical Identifiers
| 641-74-7 | |
| 146.14 | |
| KLDXJTOLSGUMSJ-UHFFFAOYNA-N | |
| 12304540 | |
| OC1COC2C(O)COC12 |
| C6H10O4 | |
| MFCD10698761,MFCD32202826,MFCD10698761 | |
| 1,4:3,6-Dianhydromannitol | |
| hexahydrofuro[3,2-b]furan-3,6-diol |
Safety and Handling
TSCA : No