missing translation for 'onlineSavingsMsg'
Learn More

Isomannide 98.0+%, TCI America™

Catalog No. p-7131391
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04065G 5 g
I040625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I04065G Supplier TCI America Supplier No. I04065G
Only null left

Chemical Identifiers

CAS 641-74-7
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.14
MDL Number MFCD10698761,MFCD32202826,MFCD10698761
InChI Key KLDXJTOLSGUMSJ-UHFFFAOYNA-N
Synonym 1,4:3,6-Dianhydromannitol
PubChem CID 12304540
IUPAC Name hexahydrofuro[3,2-b]furan-3,6-diol
SMILES OC1COC2C(O)COC12

Specifications

Melting Point 86°C
Boiling Point 150°C
Quantity 5 g
Formula Weight 146.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Isomannide