missing translation for 'onlineSavingsMsg'
Learn More

Isoliquiritigenin 97.0+%, TCI America™

Catalog No. I0822100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0822100MG 100 mg
I08221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0822100MG Supplier TCI America Supplier No. I0822100MG

Chemical Identifiers

CAS 961-29-5
Molecular Formula C15H12O4
Molecular Weight (g/mol) 256.257
MDL Number MFCD00075907
InChI Key DXDRHHKMWQZJHT-FPYGCLRLSA-N
Synonym isoliquiritigenin, 2',4,4'-trihydroxychalcone, 4,2',4'-trihydroxychalcone, 6'-deoxychalcone, e-1-2,4-dihydroxyphenyl-3-4-hydroxyphenyl prop-2-en-1-one, 2',4',4-trihydroxychalcone, isoliquirtigenin, unii-b9cti9gb8f, ccris 7676, chalcone, 2',4,4'-trihydroxy
PubChem CID 638278
ChEBI CHEBI:310312
IUPAC Name (E)-1-(2,4-dihydroxyphenyl)-3-(4-hydroxyphenyl)prop-2-en-1-one
SMILES C1=CC(=CC=C1C=CC(=O)C2=C(C=C(C=C2)O)O)O

Specifications

Melting Point 198°C
Quantity 100 mg
Formula Weight 256.26
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Isoliquiritigenin
RTECSNumber FL7080000
TSCA No
Recommended Storage Refrigerator