missing translation for 'onlineSavingsMsg'
Learn More

Isoliquiritigenin, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015974
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015974 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015974 Supplier Merck Emd Millipore Supplier No. 3898650MG

Specifications

CAS 961-29-5
Linear Formula C15H12O4
MDL Number MFCD00075907
Synonym (E)-1-(2,4-Dihydroxyphenyl)-3-(4-hydroxyphenyl)-2-propen-1-one; 4,2 ′,4 ′-Trihydroxychalcone
Type Analytical Standard
Quantity 50 mg
Formula Weight 256.25
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard