missing translation for 'onlineSavingsMsg'
Learn More

Medisca Isoleucine (L), USP

Catalog No. 14185624
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14185624 500 g
14185190 100 g
14185413 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14185624 Supplier Medisca Supplier No. 035908
Get 1 Extra Chemical When You Buy 4
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Isoleucine (L), USP comes as white, practically odorless crystals. It is soluble in water and slightly soluble in hot alcohol.

Isoleucine (L), USP comes as white, practically odorless crystals. It is soluble in water and slightly soluble in hot alcohol.

Alternative name(s): L-Isoleucine, L-Ile

Specifications

Chemical Name or Material Isoleucine (L), USP
Packaging Jar, 1000mL, White, HDPE, Round, 89-400, TE Pano Neck
CAS 73-32-5
Type Drug Standard
Molecular Weight (g/mol) 131.174
Quantity 500 g
Percent Purity 98.5 to 101.5 %
Grade USP