missing translation for 'onlineSavingsMsg'
Learn More

(-)-Isoledene, ≥95.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111015970
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015970 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015970 Supplier Merck Emd Millipore Supplier No. 43201100MG
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 95910-36-4
Density 0.902 g/mL (at 20°C)
Physical Form Neat
Linear Formula C15H24
Boiling Point 95°C (5 mmHg)
Type Analytical Standard
Quantity 100 mg
Formula Weight 204.35
Percent Purity ≥95.0% (GC)
Grade Analytical Standard