missing translation for 'onlineSavingsMsg'
Learn More

Isoindoline 98.0+%, TCI America™

Catalog No. p-7131578
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07511G 1 g
I07515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I07511G Supplier TCI America Supplier No. I07511G

Chemical Identifiers

CAS 496-12-8
Molecular Formula C8H9N
Molecular Weight (g/mol) 119.167
MDL Number MFCD00605324
InChI Key GWVMLCQWXVFZCN-UHFFFAOYSA-N
Synonym isoindoline, 1h-isoindole, 2,3-dihydro, iso-indoline, 2-azaindan, 1,2-dihydroisoindole, 1,3-dihydroisoindole, dihydroisoindole, pubchem16248, acmc-1am9e
PubChem CID 422478
IUPAC Name 2,3-dihydro-1H-isoindole
SMILES C1C2=CC=CC=C2CN1

Specifications

Melting Point 17°C
Boiling Point 113°C
Quantity 1 g
Formula Weight 119.17
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Isoindoline
TSCA No
Recommended Storage Freezer