accessibility menu, dialog, popup

UserName

CAS RN 496-12-8

IUPAC Name: 2,3-dihydro-1H-isoindole
Synonyms: isoindoline
Molecular Weight (g/mol): 119.17
Molecular Formula: C8H9N
InChi Key: GWVMLCQWXVFZCN-UHFFFAOYSA-N
SMILES: C1NCC2=CC=CC=C12
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)