missing translation for 'onlineSavingsMsg'
Learn More

Isoindoline Hydrochloride 98.0+%, TCI America™

Catalog No. I09581G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09581G 1 g
I09585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I09581G Supplier TCI America Supplier No. I09581G

Chemical Identifiers

CAS 32372-82-0
Molecular Formula C8H10ClN
Molecular Weight (g/mol) 155.63
MDL Number MFCD03502377
InChI Key NOVIRODZMIZUPA-UHFFFAOYSA-N
Synonym isoindoline hydrochloride, 2,3-dihydroisoindole hydrochloride, 2,3-dihydro-1h-isoindole hydrochloride, 2,3-dihydro-1h-isoindole hcl, 1h-isoindole, 2,3-dihydro-, hydrochloride, isoindolinehydrochloride, isoindoline hcl, d0y1rn, dihydroisoindole hydrochloride, ksc491i3f
PubChem CID 12311031
IUPAC Name 2,3-dihydro-1H-isoindole hydrochloride
SMILES Cl.C1NCC2=CC=CC=C12

Specifications

Melting Point 261°C
Color White-Yellow
Quantity 1 g
Formula Weight 155.63
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Isoindoline Hydrochloride