missing translation for 'onlineSavingsMsg'
Learn More

Isoindoline hydrochloride, 97%, Thermo Scientific™

Chemical Identifiers

CAS 32372-82-0
Molecular Formula C8H10ClN
Molecular Weight (g/mol) 155.63
MDL Number MFCD03502377
InChI Key NOVIRODZMIZUPA-UHFFFAOYSA-N
Synonym isoindoline hydrochloride, 2,3-dihydroisoindole hydrochloride, 2,3-dihydro-1h-isoindole hydrochloride, 2,3-dihydro-1h-isoindole hcl, 1h-isoindole, 2,3-dihydro-, hydrochloride, isoindolinehydrochloride, isoindoline hcl, d0y1rn, dihydroisoindole hydrochloride, ksc491i3f
PubChem CID 12311031
IUPAC Name 2,3-dihydro-1H-isoindole hydrochloride
SMILES Cl.C1NCC2=CC=CC=C12
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC435850050
SDS View Documents
Thermo Scientific Chemicals
435850050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC435850250
SDS View Documents
Thermo Scientific Chemicals
435850250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

32372-82-0
155.63
NOVIRODZMIZUPA-UHFFFAOYSA-N
12311031
Cl.C1NCC2=CC=CC=C12
C8H10ClN
MFCD03502377
isoindoline hydrochloride, 2,3-dihydroisoindole hydrochloride, 2,3-dihydro-1h-isoindole hydrochloride, 2,3-dihydro-1h-isoindole hcl, 1h-isoindole, 2,3-dihydro-, hydrochloride, isoindolinehydrochloride, isoindoline hcl, d0y1rn, dihydroisoindole hydrochloride, ksc491i3f
2,3-dihydro-1H-isoindole hydrochloride

Specifications

32372-82-0
Authentic
Glass bottle
MFCD03502377
isoindoline hydrochloride, 2,3-dihydroisoindole hydrochloride, 2,3-dihydro-1h-isoindole hydrochloride, 2,3-dihydro-1h-isoindole hcl, 1h-isoindole, 2,3-dihydro-, hydrochloride, isoindolinehydrochloride, isoindoline hcl, d0y1rn, dihydroisoindole hydrochloride, ksc491i3f
Cl.C1NCC2=CC=CC=C12
155.63
155.63
Isoindoline hydrochloride
59.27 to 64.21% C, 8.64 to 9.36% N
96% min. (HPLC)
C8H10ClN
5g
NOVIRODZMIZUPA-UHFFFAOYSA-N
2,3-dihydro-1H-isoindole hydrochloride
12311031
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only