missing translation for 'onlineSavingsMsg'
Learn More

Isoindoline hydrochloride, 97%

Catalog No. AC435850050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC435850050 5 g Glass bottle
AC435850250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC435850050 Supplier Thermo Scientific Chemicals Supplier No. 435850050

CAS: 32372-82-0 | C8H10ClN | 155.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 32372-82-0
Molecular Formula C8H10ClN
Molecular Weight (g/mol) 155.63
MDL Number MFCD03502377
InChI Key NOVIRODZMIZUPA-UHFFFAOYSA-N
Synonym isoindoline hydrochloride, 2,3-dihydroisoindole hydrochloride, 2,3-dihydro-1h-isoindole hydrochloride, 2,3-dihydro-1h-isoindole hcl, 1h-isoindole, 2,3-dihydro-, hydrochloride, isoindolinehydrochloride, isoindoline hcl, d0y1rn, dihydroisoindole hydrochloride, ksc491i3f
PubChem CID 12311031
SMILES Cl.C1NCC2=CC=CC=C12

Specifications

Melting Point 255°C to 256°C
Color White
Elemental Analysis 59.27 to 64.21% C, 8.64 to 9.36% N
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
IUPAC Name 2,3-dihydro-1H-isoindole hydrochloride
Formula Weight 155.63
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Isoindoline hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only