missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isoindolin-1-one 98.0+%, TCI America™
$150.98 - $508.89
Chemical Identifiers
| CAS | 480-91-1 |
|---|---|
| Molecular Formula | C8H7NO |
| Molecular Weight (g/mol) | 133.15 |
| MDL Number | MFCD03085939 |
| InChI Key | PXZQEOJJUGGUIB-UHFFFAOYSA-N |
| Synonym | 2,3-Dihydro-1H-isoindol-1-one |
| PubChem CID | 10199 |
| ChEBI | CHEBI:74235 |
| IUPAC Name | 2,3-dihydroisoindol-1-one |
| SMILES | C1C2=CC=CC=C2C(=O)N1 |
Chemical Identifiers
| 480-91-1 | |
| 133.15 | |
| PXZQEOJJUGGUIB-UHFFFAOYSA-N | |
| 10199 | |
| 2,3-dihydroisoindol-1-one |
| C8H7NO | |
| MFCD03085939 | |
| 2,3-Dihydro-1H-isoindol-1-one | |
| CHEBI:74235 | |
| C1C2=CC=CC=C2C(=O)N1 |
Specifications
| 480-91-1 | |
| White-Yellow | |
| C8H7NO | |
| 1 g | |
| PXZQEOJJUGGUIB-UHFFFAOYSA-N | |
| 2,3-dihydroisoindol-1-one | |
| 10199 | |
| 133.15 | |
| Crystalline Powder |
| 151°C | |
| 337°C | |
| MFCD03085939 | |
| 2,3-Dihydro-1H-isoindol-1-one | |
| C1C2=CC=CC=C2C(=O)N1 | |
| 133.15 | |
| CHEBI:74235 | |
| ≥98.0% (GC) | |
| Isoindolin-1-one |
Safety and Handling
TSCA : No