missing translation for 'onlineSavingsMsg'
Learn More

Isoindolin-1-one 98.0+%, TCI America™

Catalog No. I10641G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I10641G 1 g
I10645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I10641G Supplier TCI America Supplier No. I10641G

Chemical Identifiers

CAS 480-91-1
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD03085939
InChI Key PXZQEOJJUGGUIB-UHFFFAOYSA-N
Synonym 2,3-Dihydro-1H-isoindol-1-one
PubChem CID 10199
ChEBI CHEBI:74235
IUPAC Name 2,3-dihydroisoindol-1-one
SMILES C1C2=CC=CC=C2C(=O)N1

Specifications

Melting Point 151°C
Color White-Yellow
Boiling Point 337°C
Quantity 1 g
Formula Weight 133.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Isoindolin-1-one