Learn More
Isododecane, mixture of isomers, tech. 80%
CAS: 31807-55-3 | C12H26 | 170.34 g/mol
Supplier: Thermo Scientific Chemicals A142350F
Description
Isododecane, mixture of isomer acts as a reaction medium used for polymerization reactions. It is also useful as an emollient and solvent in skin care products due to its high spreadability, low viscosity and density. It is commonly used in anti-aging serums and also useful in many different cosmetic items like eyeliner, hair care, hair sprays, perfume, conditioners and lotions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIsododecane, mixture of isomer acts as a reaction medium used for polymerization reactions. It is also useful as an emollient and solvent in skin care products due to its high spreadability, low viscosity and density. It is commonly used in anti-aging serums and also useful in many different cosmetic items like eyeliner, hair care, hair sprays, perfume, conditioners and lotions.
Solubility
Immiscible with water.
Notes
Incompatible with oxidizing agents.
Specifications
| Isododecane | |
| 31807-55-3 | |
| 170°C to 195°C | |
| 80% | |
| 1.421 | |
| 2500 mL | |
| isododecane, undecane, 2-methyl, 2-methylhendecane, unii-h40fl8477b, isodo-decane, acmc-1bc2q, isododecane, mixture of isomers | |
| GTJOHISYCKPIMT-UHFFFAOYSA-N | |
| 2-methylundecane | |
| 23459 | |
| 80% | |
| Liquid |
| mixture of isomers | |
| 0.74 | |
| 42°C (108°F) | |
| C12H26 | |
| MFCD00036106 | |
| UN3295 | |
| Immiscible with water. | |
| CCCCCCCCCC(C)C | |
| 170.34 | |
| 170.34 | |
| Technical |
Chemical Identifiers
| 31807-55-3 | |
| 170.34 | |
| GTJOHISYCKPIMT-UHFFFAOYSA-N | |
| 23459 | |
| CCCCCCCCCC(C)C |
| C12H26 | |
| MFCD00036106 | |
| isododecane, undecane, 2-methyl, 2-methylhendecane, unii-h40fl8477b, isodo-decane, acmc-1bc2q, isododecane, mixture of isomers | |
| 2-methylundecane |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c
H226-H304
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber : 250-816-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only