missing translation for 'onlineSavingsMsg'
Learn More

Isobutyryl-L-carnitine, ≥97.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013743
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013743 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013743 Supplier Merck Emd Millipore Supplier No. 9129910MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 25518-49-4
Linear Formula C11H21 NO4
MDL Number MFCD01674829
Synonym (2 R)-3-Carboxy-N,N,N-trimethyl-2-(2-methyl-1-oxopropoxy)-1-propanaminium inner salt; L-Carnitine isobutyryl ester; iC4-Carnitine
Type Analytical Standard
Quantity 10 mg
Formula Weight 231.29
Percent Purity ≥97.0% (HPLC)
Grade Analytical Standard