missing translation for 'onlineSavingsMsg'
Learn More

Isobutyryl chloride, 98%

CAS: 79-30-1 | C4H7ClO | 106.55 g/mol

$57.96 - $57.96

Chemical Identifiers

CAS 79-30-1
Molecular Formula C4H7ClO
Molecular Weight (g/mol) 106.55
MDL Number MFCD00000717
InChI Key DGMOBVGABMBZSB-UHFFFAOYSA-N
Synonym isobutyryl chloride, 2-methylpropionyl chloride, propanoyl chloride, 2-methyl, isobutanoyl chloride, isobutyric acid chloride, isobutyroyl chloride, isobutyrylchloride, dimethylacetyl chloride, chloro isopropyl ketone, alpha-methylpropionyl chloride
PubChem CID 62325
IUPAC Name 2-methylpropanoyl chloride
SMILES CC(C)C(=O)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Qty  
Catalog Number Mfr. No. Quantity Packaging Price Qty  
AC160660100
SDS View Documents
Thermo Scientific Chemicals
160660100
10 mL Glass bottle
Each for $57.96
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

79-30-1
106.55
DGMOBVGABMBZSB-UHFFFAOYSA-N
62325
CC(C)C(=O)Cl
C4H7ClO
MFCD00000717
isobutyryl chloride, 2-methylpropionyl chloride, propanoyl chloride, 2-methyl, isobutanoyl chloride, isobutyric acid chloride, isobutyroyl chloride, isobutyrylchloride, dimethylacetyl chloride, chloro isopropyl ketone, alpha-methylpropionyl chloride
2-methylpropanoyl chloride

Specifications

79-30-1
1.0170g/mL
1°C
97.5% min. (GC)
C4H7ClO
(CH3)2CHCOCl
10 mL
1.017
Solubility in water: reacts. Other solubilities: reacts with alcohol,soluble in ether
CC(C)C(=O)Cl
106.55
0.55 mPa.s (20°C)
98%
Isobutyryl chloride, 98%
-90.0°C
91.0°C to 93.0°C
Authentic
Glass bottle
1.4060 to 1.4080
MFCD00000717
02, 293
isobutyryl chloride, 2-methylpropionyl chloride, propanoyl chloride, 2-methyl, isobutanoyl chloride, isobutyric acid chloride, isobutyroyl chloride, isobutyrylchloride, dimethylacetyl chloride, chloro isopropyl ketone, alpha-methylpropionyl chloride
DGMOBVGABMBZSB-UHFFFAOYSA-N
2-methylpropanoyl chloride
62325
106.55
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Harmful if swallowed.
Fatal if inhaled.
May be corrosive to metals.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 201-194-1

RTECSNumber : UC3944000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only