accessibility menu, dialog, popup

UserName

H3CCH3ClO

CAS RN 79-30-1

IUPAC Name: 2-methylpropanoyl chloride
Synonyms: propanoyl chloride, 2-methyl-
Molecular Weight (g/mol): 106.55
Molecular Formula: C4H7ClO
InChi Key: DGMOBVGABMBZSB-UHFFFAOYSA-N
SMILES: CC(C)C(Cl)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)