missing translation for 'onlineSavingsMsg'
Learn More

Isobutyryl chloride, 98%

Catalog No. AC160660100
Change view
Click to view available options
Quantity:
10 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC160660100 10 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC160660100 Supplier Thermo Scientific Chemicals Supplier No. 160660100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 79-30-1 | C4H7ClO | 106.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79-30-1
Molecular Formula C4H7ClO
Molecular Weight (g/mol) 106.55
MDL Number MFCD00000717
InChI Key DGMOBVGABMBZSB-UHFFFAOYSA-N
Synonym isobutyryl chloride, 2-methylpropionyl chloride, propanoyl chloride, 2-methyl, isobutanoyl chloride, isobutyric acid chloride, isobutyroyl chloride, isobutyrylchloride, dimethylacetyl chloride, chloro isopropyl ketone, alpha-methylpropionyl chloride
PubChem CID 62325
IUPAC Name 2-methylpropanoyl chloride
SMILES CC(C)C(=O)Cl

Specifications

Melting Point -90.0°C
Density 1.0170g/mL
Boiling Point 91.0°C to 93.0°C
Flash Point 1°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4060 to 1.4080
Linear Formula (CH3)2CHCOCl
Quantity 10 mL
Beilstein 02, 293
Specific Gravity 1.017
Solubility Information Solubility in water: reacts. Other solubilities: reacts with alcohol,soluble in ether
Viscosity 0.55 mPa.s (20°C)
Formula Weight 106.55
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Isobutyryl chloride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Harmful if swallowed.
Fatal if inhaled.
May be corrosive to metals.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 201-194-1
RTECSNumber UC3944000
TSCA TSCA

RUO – Research Use Only