missing translation for 'onlineSavingsMsg'
Learn More

Isobutyronitrile 98.0+%, TCI America™

Catalog No. p-7131219
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I011225ML 25 mL
I0112500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I011225ML Supplier TCI America Supplier No. I011225ML
Only null left

Chemical Identifiers

CAS 78-82-0
Molecular Formula C4H7N
Molecular Weight (g/mol) 69.107
MDL Number MFCD00001873
InChI Key LRDFRRGEGBBSRN-UHFFFAOYSA-N
Synonym isobutyronitrile, isopropyl cyanide, propanenitrile, 2-methyl, 2-methylpropionitrile, isopropylnitrile, 2-cyanopropane, dimethylacetonitrile, isopropyl nitrile, 1-cyano-1-methylethane, isopropylkyanid
PubChem CID 6559
ChEBI CHEBI:28638
IUPAC Name 2-methylpropanenitrile
SMILES CC(C)C#N

Specifications

Melting Point -72°C
Color Colorless
Boiling Point 108°C
Quantity 25 mL
UN Number 2284
Formula Weight 69.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutyronitrile
EINECSNumber (2)-1510
RTECSNumber TZ4900000
TSCA Yes