missing translation for 'onlineSavingsMsg'
Learn More

Isobutyronitrile, 99%, Spectrum™ Chemical

Catalog No. 18603511
Change view
Click to view available options
Quantity:
100 mL
200 L
Packaging:
Amber Glass Bottle
Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603511 100 mL Amber Glass Bottle
18603513 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18603511 Supplier Spectrum Chemical Mfg Cor Supplier No. I2199100ML

I2199, 78-82-0, C4H7N

Spectrum™ Chemical Isobutyronitrile

Chemical Identifiers

CAS 78-82-0
Molecular Formula C4H7N
Molecular Weight (g/mol) 69.11
InChI Key LRDFRRGEGBBSRN-UHFFFAOYSA-N
IUPAC Name 2-methylpropanenitrile
SMILES CC(C)C#N

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 100 mL
UN Number UN2284
Percent Purity 99%
Grade Reagent