missing translation for 'onlineSavingsMsg'
Learn More

Isobutyric Anhydride 95.0+%, TCI America™

Catalog No. p-7131218
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
I011125ML 25 mL
I0111500ML 500 mL
2 options

Catalog No. I011125ML

Supplier: TCI America I011125ML

Only null left

Chemical Identifiers

CAS 97-72-3
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.197
MDL Number MFCD00008913
InChI Key LSACYLWPPQLVSM-UHFFFAOYSA-N
Synonym isobutyric anhydride, isobutyric acid anhydride, propanoic acid, 2-methyl-, anhydride, 2-methylpropanoic anhydride, isobutyricanhydride, isobutyryl anhydride, 2-methylpropionic anhydride, unii-n85a80fjdt, n85a80fjdt, 2-methylpropanoic acid anhydride
PubChem CID 7346
ChEBI CHEBI:84261
IUPAC Name 2-methylpropanoyl 2-methylpropanoate
SMILES CC(C)C(=O)OC(=O)C(C)C

Specifications

Color Yellow
Boiling Point 182°C
Quantity 25 mL
UN Number 3265
Formula Weight 158.2
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Isobutyric Anhydride
EINECSNumber (2)-4182
TSCA Yes