missing translation for 'onlineSavingsMsg'
Learn More

Isobutyric acid, MilliporeSigma™ Supelco™

Catalog No. 111020663
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020663 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020663 Supplier Merck Emd Millipore Supplier No. 46935U
Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Packaging Ampule
Ignition Point 824°F
CAS 79-31-2
Vapor Pressure 1.5 mmHg (20°C)
Vapor Density 3.04 (vs. air)
Density 0.95 g/mL (at 25°C)
Flash Point 55°C
Physical Form Neat
Linear Formula (CH3)2 CHCO2 H
MDL Number MFCD00002658
Melting Point -47°C
Boiling Point 153°C to 154°C
Synonym 2-Methylpropionic acid
Type Food and Cosmetic Component Standard
Quantity 500 mg
Formula Weight 88.11
Grade Analytical Standard
Show More Show Less