missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isobutyraldehyde 98.0+%, TCI America™
$33.56 - $50.33
Chemical Identifiers
| CAS | 78-84-2 |
|---|---|
| Molecular Formula | C4H8O |
| Molecular Weight (g/mol) | 72.11 |
| MDL Number | MFCD00006980 |
| InChI Key | AMIMRNSIRUDHCM-UHFFFAOYSA-N |
| Synonym | isobutyraldehyde, isobutanal, isobutylaldehyde, propanal, 2-methyl, 2-methylpropionaldehyde, isobutyric aldehyde, valine aldehyde, isopropylaldehyde, 2-methyl-1-propanal, isobutaldehyde |
| PubChem CID | 6561 |
| ChEBI | CHEBI:48943 |
| IUPAC Name | 2-methylpropanal |
| SMILES | CC(C)C=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
I010125ML
|
TCI America
I010125ML |
25 mL |
Each for $33.56
|
|
|||||
|
I0101500ML
|
TCI America
I0101500ML |
500 mL |
Each for $50.33
|
|
|||||
Chemical Identifiers
| 78-84-2 | |
| 72.11 | |
| AMIMRNSIRUDHCM-UHFFFAOYSA-N | |
| 6561 | |
| 2-methylpropanal |
| C4H8O | |
| MFCD00006980 | |
| isobutyraldehyde, isobutanal, isobutylaldehyde, propanal, 2-methyl, 2-methylpropionaldehyde, isobutyric aldehyde, valine aldehyde, isopropylaldehyde, 2-methyl-1-propanal, isobutaldehyde | |
| CHEBI:48943 | |
| CC(C)C=O |
Specifications
| 78-84-2 | |
| Yellow | |
| C4H8O | |
| 25 mL | |
| isobutyraldehyde, isobutanal, isobutylaldehyde, propanal, 2-methyl, 2-methylpropionaldehyde, isobutyric aldehyde, valine aldehyde, isopropylaldehyde, 2-methyl-1-propanal, isobutaldehyde | |
| CC(C)C=O | |
| 72.11 | |
| CHEBI:48943 | |
| ≥98.0% (GC) | |
| Isobutyraldehyde |
| -65°C | |
| 65°C | |
| MFCD00006980 | |
| 2045 | |
| AMIMRNSIRUDHCM-UHFFFAOYSA-N | |
| 2-methylpropanal | |
| 6561 | |
| 72.11 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0494
RTECSNumber : NQ4025000
TSCA : Yes
Recommended Storage : Refrigerator