missing translation for 'onlineSavingsMsg'
Learn More

Isobutyraldehyde 98.0+%, TCI America™

Catalog No. I010125ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I010125ML 25 mL
I0101500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I010125ML Supplier TCI America Supplier No. I010125ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 78-84-2
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00006980
InChI Key AMIMRNSIRUDHCM-UHFFFAOYSA-N
Synonym isobutyraldehyde, isobutanal, isobutylaldehyde, propanal, 2-methyl, 2-methylpropionaldehyde, isobutyric aldehyde, valine aldehyde, isopropylaldehyde, 2-methyl-1-propanal, isobutaldehyde
PubChem CID 6561
ChEBI CHEBI:48943
IUPAC Name 2-methylpropanal
SMILES CC(C)C=O

Specifications

Melting Point -65°C
Color Yellow
Boiling Point 65°C
Quantity 25 mL
UN Number 2045
Formula Weight 72.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutyraldehyde
EINECSNumber (2)-0494
RTECSNumber NQ4025000
TSCA Yes
Recommended Storage Refrigerator