missing translation for 'onlineSavingsMsg'
Learn More

Isobutyraldehyde Diethyl Acetal 96.0+%, TCI America™

Catalog No. I02875ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I02875ML 5 mL
I028725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I02875ML Supplier TCI America Supplier No. I02875ML

Chemical Identifiers

CAS 1741-41-9
Molecular Formula C8H18O2
Molecular Weight (g/mol) 146.23
MDL Number MFCD00059384
InChI Key KZDFOVZPOBSHDH-UHFFFAOYSA-N
Synonym 1,1-Diethoxyisobutane
PubChem CID 519415
IUPAC Name 1,1-diethoxy-2-methylpropane
SMILES CCOC(C(C)C)OCC

Specifications

Color Colorless
Boiling Point 138°C
Quantity 5 mL
UN Number 1993
Formula Weight 146.23
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutyraldehyde Diethyl Acetal