accessibility menu, dialog, popup

UserName

CAS RN 1741-41-9

H3COOCH3CH3H3C

CAS RN 1741-41-9

IUPAC Name: 1,1-diethoxy-2-methylpropane
Synonyms: 1,1-diethoxy-2-methylpropane
Molecular Weight (g/mol): 146.23
Molecular Formula: C8H18O2
InChi Key: KZDFOVZPOBSHDH-UHFFFAOYSA-N
SMILES: CCOC(OCC)C(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results