missing translation for 'onlineSavingsMsg'
Learn More

Isobutyraldehyde, 99+%

Catalog No. p-3737951
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC122510010 1 L Glass bottle
AC122510025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC122510010 Supplier Thermo Scientific Chemicals Supplier No. 12251 0010
Only null left

CAS: 78-84-2 | C4H8O | 72.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 78-84-2
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00006980
InChI Key AMIMRNSIRUDHCM-UHFFFAOYSA-N
Synonym isobutyraldehyde, isobutanal, isobutylaldehyde, propanal, 2-methyl, 2-methylpropionaldehyde, isobutyric aldehyde, valine aldehyde, isopropylaldehyde, 2-methyl-1-propanal, isobutaldehyde
PubChem CID 6561
ChEBI CHEBI:48943
IUPAC Name 2-methylpropanal
SMILES CC(C)C=O

Specifications

Melting Point -65.0°C
Density 0.7900g/mL
Boiling Point 63.0°C
Flash Point −24°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.3710 to 1.3740
Linear Formula (CH3)2CHCHO
Quantity 1 L
Beilstein 01, 671
Specific Gravity 0.79
Merck Index 15, 5199
Solubility Information Solubility in water: 75g/L (20°C). Other solubilities: miscible with ethanol, ether, carbon disulfide,, acetone, benzene, toluene and chloroform
Viscosity 0.45 mPa.s (20°C)
Formula Weight 72.11
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Isobutyraldehyde
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond conta
EINECSNumber 201-149-6

RUO – Research Use Only