missing translation for 'onlineSavingsMsg'
Learn More

Isobutylene Sulfide 98.0+%, TCI America™

Catalog No. I04615ML
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
5 mL
Catalog No. Quantity
I04615ML 5 mL
I046125ML 25 mL
2 options

Catalog No. I04615ML

Supplier: TCI America I04615ML

Only null left

Chemical Identifiers

CAS 3772-13-2
Molecular Formula C4H8S
Molecular Weight (g/mol) 88.17
MDL Number MFCD00191662
InChI Key HGJOFJDIHKHKAU-UHFFFAOYSA-N
Synonym 2,2-Dimethylethylene Sulfide, 2,2-Dimethylthiirane, 2-Methylpropylene Sulfide
PubChem CID 138050
IUPAC Name 2,2-dimethylthiirane
SMILES CC1(C)CS1

Specifications

Color Yellow
Boiling Point 86°C
Quantity 5 mL
UN Number 1993
Formula Weight 88.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutylene Sulfide
TSCA No
Recommended Storage Refrigerator