missing translation for 'onlineSavingsMsg'
Learn More

Isobutylene Oxide 98.0+%, TCI America™

Catalog No. I032325ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I032325ML 25 mL
I0323500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I032325ML Supplier TCI America Supplier No. I032325ML

Chemical Identifiers

CAS 558-30-5
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.107
MDL Number MFCD00066354
InChI Key GELKGHVAFRCJNA-UHFFFAOYSA-N
Synonym isobutylene oxide, 1,2-epoxy-2-methylpropane, isobutene oxide, isobutylene epoxide, isobutyleneoxide, oxirane, 2,2-dimethyl, 1,2-epoxyisobutane, 1,1-dimethylethylene oxide, 1,2-isobutylene oxide, 2-methyl-1-propene oxide
PubChem CID 11208
IUPAC Name 2,2-dimethyloxirane
SMILES CC1(CO1)C

Specifications

Color Colorless
pH 6.7
Boiling Point 52°C
Quantity 25 mL
UN Number 1993
Formula Weight 72.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutylene Oxide
EINECSNumber (2)-0229
RTECSNumber TZ3580000
TSCA Yes