accessibility menu, dialog, popup

UserName

CAS RN 558-30-5

IUPAC Name: 2,2-dimethyloxirane
Synonyms: 2,2-dimethyloxirane
Molecular Weight (g/mol): 72.11
Molecular Formula: C4H8O
InChi Key: GELKGHVAFRCJNA-UHFFFAOYSA-N
SMILES: CC1(C)CO1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)