missing translation for 'onlineSavingsMsg'
Learn More

Isobutylene oxide, 98%

CAS: 558-30-5 | C4H8O | 72.11 g/mol

$275.80 - $275.80

Chemical Identifiers

CAS 558-30-5
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00066354
InChI Key GELKGHVAFRCJNA-UHFFFAOYSA-N
Synonym isobutylene oxide, 1,2-epoxy-2-methylpropane, isobutene oxide, isobutylene epoxide, isobutyleneoxide, oxirane, 2,2-dimethyl, 1,2-epoxyisobutane, 1,1-dimethylethylene oxide, 1,2-isobutylene oxide, 2-methyl-1-propene oxide
PubChem CID 11208
IUPAC Name 2,2-dimethyloxirane
SMILES CC1(CO1)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC348500250
SDS View Documents
Thermo Scientific Chemicals
348500250
25 mL Glass bottle
Each for $275.80
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

558-30-5
72.11
GELKGHVAFRCJNA-UHFFFAOYSA-N
11208
CC1(CO1)C
C4H8O
MFCD00066354
isobutylene oxide, 1,2-epoxy-2-methylpropane, isobutene oxide, isobutylene epoxide, isobutyleneoxide, oxirane, 2,2-dimethyl, 1,2-epoxyisobutane, 1,1-dimethylethylene oxide, 1,2-isobutylene oxide, 2-methyl-1-propene oxide
2,2-dimethyloxirane

Specifications

558-30-5
0.8080g/mL
−32°C
97.5% min. (GC)
C4H8O
MFCD00066354
17, V,1, 58
isobutylene oxide, 1,2-epoxy-2-methylpropane, isobutene oxide, isobutylene epoxide, isobutyleneoxide, oxirane, 2,2-dimethyl, 1,2-epoxyisobutane, 1,1-dimethylethylene oxide, 1,2-isobutylene oxide, 2-methyl-1-propene oxide
GELKGHVAFRCJNA-UHFFFAOYSA-N
2,2-dimethyloxirane
11208
72.11
Liquid
Colorless
51.0°C to 52.0°C
Authentic
Glass bottle
1.3710 to 1.3730
25 mL
0.808
Solubility in water: 30g/L (20°C). Other solubilities: soluble in ethanol,diethyl ether
CC1(CO1)C
72.11
0.513 mm2/s (20°C)
98%
Isobutylene oxide, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Do not breat

GHS Signal Word: Danger

EINECSNumber : 209-193-8

RTECSNumber : TZ3580000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only