missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isobutylbenzene 99.0+%, TCI America™
$59.69 - $283.00
Chemical Identifiers
| CAS | 538-93-2 |
|---|---|
| Molecular Formula | C10H14 |
| Molecular Weight (g/mol) | 134.22 |
| MDL Number | MFCD00008936 |
| InChI Key | KXUHSQYYJYAXGZ-UHFFFAOYSA-N |
| Synonym | isobutylbenzene, benzene, 2-methylpropyl, 2-methylpropyl benzene, benzene, isobutyl, 1-phenyl-2-methylpropane, 2-methyl-1-phenylpropane, 1-phenylisobutane, 2-phenyl-2-methylpropane, iso-butylbenzene, 1-isobutylbenzene |
| PubChem CID | 10870 |
| ChEBI | CHEBI:43261 |
| IUPAC Name | (2-methylpropyl)benzene |
| SMILES | CC(C)CC1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
I009725ML
|
TCI America
I009725ML |
25 mL |
Each for $59.69
|
|
|||||
|
I0097500ML
|
TCI America
I0097500ML |
500 mL |
Each for $283.00
|
|
|||||
Chemical Identifiers
| 538-93-2 | |
| 134.22 | |
| KXUHSQYYJYAXGZ-UHFFFAOYSA-N | |
| 10870 | |
| (2-methylpropyl)benzene |
| C10H14 | |
| MFCD00008936 | |
| isobutylbenzene, benzene, 2-methylpropyl, 2-methylpropyl benzene, benzene, isobutyl, 1-phenyl-2-methylpropane, 2-methyl-1-phenylpropane, 1-phenylisobutane, 2-phenyl-2-methylpropane, iso-butylbenzene, 1-isobutylbenzene | |
| CHEBI:43261 | |
| CC(C)CC1=CC=CC=C1 |
Specifications
| 538-93-2 | |
| Colorless | |
| C10H14 | |
| 25 mL | |
| isobutylbenzene, benzene, 2-methylpropyl, 2-methylpropyl benzene, benzene, isobutyl, 1-phenyl-2-methylpropane, 2-methyl-1-phenylpropane, 1-phenylisobutane, 2-phenyl-2-methylpropane, iso-butylbenzene, 1-isobutylbenzene | |
| CC(C)CC1=CC=CC=C1 | |
| 134.22 | |
| CHEBI:43261 | |
| ≥99.0% (GC) | |
| Isobutylbenzene |
| -52°C | |
| 172°C | |
| MFCD00008936 | |
| 2709 | |
| KXUHSQYYJYAXGZ-UHFFFAOYSA-N | |
| (2-methylpropyl)benzene | |
| 10870 | |
| 134.22 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-0022
RTECSNumber : DA3550000
TSCA : Yes