missing translation for 'onlineSavingsMsg'
Learn More

Isobutylbenzene 99.0+%, TCI America™

Catalog No. I0097500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0097500ML 500 mL
I009725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0097500ML Supplier TCI America Supplier No. I0097500ML

Chemical Identifiers

CAS 538-93-2
Molecular Formula C10H14
Molecular Weight (g/mol) 134.22
MDL Number MFCD00008936
InChI Key KXUHSQYYJYAXGZ-UHFFFAOYSA-N
Synonym isobutylbenzene, benzene, 2-methylpropyl, 2-methylpropyl benzene, benzene, isobutyl, 1-phenyl-2-methylpropane, 2-methyl-1-phenylpropane, 1-phenylisobutane, 2-phenyl-2-methylpropane, iso-butylbenzene, 1-isobutylbenzene
PubChem CID 10870
ChEBI CHEBI:43261
IUPAC Name (2-methylpropyl)benzene
SMILES CC(C)CC1=CC=CC=C1

Specifications

Melting Point -52°C
Color Colorless
Boiling Point 172°C
Quantity 500 mL
UN Number 2709
Formula Weight 134.22
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutylbenzene
EINECSNumber (3)-0022
RTECSNumber DA3550000
TSCA Yes