missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isobutylbenzene 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | -52°C |
| Color | Colorless |
| Boiling Point | 172°C |
| Quantity | 25 mL |
| UN Number | 2709 |
| Formula Weight | 134.22 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Isobutylbenzene |
Safety and Handling
| EINECSNumber | (3)-0022 |
| RTECSNumber | DA3550000 |
| TSCA | Yes |