missing translation for 'onlineSavingsMsg'
Learn More

Isobutylamine Hydroiodide 97.0+%, TCI America™

Catalog No. I09355G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09355G 5 g
I09351G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I09355G Supplier TCI America Supplier No. I09355G

Chemical Identifiers

CAS 205508-75-4
Molecular Formula C4H12IN
Molecular Weight (g/mol) 201.051
InChI Key FCTHQYIDLRRROX-UHFFFAOYSA-N
Synonym Isobutylammonium Iodide
PubChem CID 88371729
IUPAC Name 2-methylpropan-1-amine;hydroiodide
SMILES CC(C)CN.I

Specifications

Color White
Quantity 5 g
Formula Weight 201.05
Percent Purity ≥97.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Isobutylamine Hydroiodide