accessibility menu, dialog, popup

UserName

CH3H3CH2NHI

CAS RN 205508-75-4

IUPAC Name: 2-methylpropan-1-amine hydroiodide
Synonyms: isobutylamine hydroiodide
Molecular Weight (g/mol): 201.05
Molecular Formula: C4H12IN
InChi Key: FCTHQYIDLRRROX-UHFFFAOYSA-N
SMILES: I.CC(C)CN
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results