missing translation for 'onlineSavingsMsg'
Learn More

Isobutylamine Hydrobromide 98.0+%, TCI America™

Catalog No. I10075G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I10075G 5 g
I10071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I10075G Supplier TCI America Supplier No. I10075G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 74098-36-5
Molecular Formula C4H12BrN
Molecular Weight (g/mol) 154.05
MDL Number MFCD29089367
InChI Key RFYSBVUZWGEPBE-UHFFFAOYSA-N
Synonym Isobutylammonium Bromide
PubChem CID 85987475
IUPAC Name 2-methylpropan-1-amine hydrobromide
SMILES Br.CC(C)CN

Specifications

Melting Point 182°C
Color White
Quantity 5 g
Formula Weight 154.05
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Isobutylamine Hydrobromide