accessibility menu, dialog, popup

UserName

CAS RN 74098-36-5

IUPAC Name: 2-methylpropan-1-amine hydrobromide
Synonyms: isobutylamine hydrobromide
Molecular Weight (g/mol): 154.05
Molecular Formula: C4H12BrN
InChi Key: RFYSBVUZWGEPBE-UHFFFAOYSA-N
SMILES: Br.CC(C)CN
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results