missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl Tiglate (stabilized with HQ) 96.0+%, TCI America™

Catalog No. T127425ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T127425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T127425ML Supplier TCI America Supplier No. T127425ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 61692-84-0
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.23
MDL Number MFCD00016629
InChI Key XDEGQMQKHFPBEW-VMPITWQZSA-N
Synonym Tiglic Acid Isobutyl Ester
PubChem CID 5367806
IUPAC Name 2-methylpropyl (2E)-2-methylbut-2-enoate
SMILES C\C=C(/C)C(=O)OCC(C)C

Specifications

Color Yellow
Quantity 25 mL
Formula Weight 156.23
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutyl Tiglate (stabilized with HQ)
EINECSNumber (2)-0969
TSCA Yes