accessibility menu, dialog, popup

UserName

H3CCH3OOCH3CH3(E)

CAS RN 61692-84-0

IUPAC Name: 2-methylpropyl (2E)-2-methylbut-2-enoate
Synonyms: isobutyl tiglate
Molecular Weight (g/mol): 156.23
Molecular Formula: C9H16O2
InChi Key: XDEGQMQKHFPBEW-VMPITWQZSA-N
SMILES: C\C=C(/C)C(=O)OCC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results