missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl Salicylate 99.0+%, TCI America™

Catalog No. p-7135763
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S001325G 25 g
S0013500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S001325G Supplier TCI America Supplier No. S001325G
Only null left

Chemical Identifiers

CAS 87-19-4
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00020035
InChI Key PTXDBYSCVQQBNF-UHFFFAOYSA-N
Synonym 2-Hydroxybenzoic Acid Isobutyl Ester, Salicylic Acid Isobutyl Ester
PubChem CID 6873
IUPAC Name 2-methylpropyl 2-hydroxybenzoate
SMILES CC(C)COC(=O)C1=CC=CC=C1O

Specifications

Color Colorless
Boiling Point 259°C
Quantity 25 g
Formula Weight 194.23
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutyl Salicylate
EINECSNumber (3)-1585
RTECSNumber VO4320000
TSCA Yes