missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl Propionate 98.0+%, TCI America™

Catalog No. p-7134646
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P050625ML 25 mL
P0506100ML 100 mL
P0506500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P050625ML Supplier TCI America Supplier No. P050625ML
Only null left

Chemical Identifiers

CAS 540-42-1
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
MDL Number MFCD00009307
InChI Key FZXRXKLUIMKDEL-UHFFFAOYSA-N
Synonym isobutyl propionate, isobutyl propanoate, 2-methylpropyl propionate, propanoic acid, 2-methylpropyl ester, propionic acid isobutyl ester, propionic acid, isobutyl ester, methyl isobutyl acetate, 2-methyl-1-propyl propanoate, unii-8q382jsy9t, isobutyl propionate natural
PubChem CID 10895
IUPAC Name 2-methylpropyl propanoate
SMILES CCC(=O)OCC(C)C

Specifications

Color Colorless
Boiling Point 138°C
Quantity 25 mL
UN Number 2394
Formula Weight 130.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutyl Propionate
EINECSNumber (2)-0774
RTECSNumber UF4930000
TSCA Yes