CAS RN 540-42-1
CAS RN 540-42-1
IUPAC Name:
2-methylpropyl propanoate
Synonyms:
isobutyl propionate
Molecular Weight (g/mol):
130.19
Molecular Formula:
C7H14O2
InChi Key:
FZXRXKLUIMKDEL-UHFFFAOYSA-N
SMILES:
CCC(=O)OCC(C)C
1
–
1
of
1
results
1
Isobutyl Propionate 98.0+%, TCI America™
CAS: 540-42-1 Molecular Formula: C7H14O2 Molecular Weight (g/mol): 130.19 MDL Number: MFCD00009307 InChI Key: FZXRXKLUIMKDEL-UHFFFAOYSA-N Synonym: isobutyl propionate,isobutyl propanoate,2-methylpropyl propionate,propanoic acid, 2-methylpropyl ester,propionic acid isobutyl ester,propionic acid, isobutyl ester,methyl isobutyl acetate,2-methyl-1-propyl propanoate,unii-8q382jsy9t,isobutyl propionate natural PubChem CID: 10895 IUPAC Name: 2-methylpropyl propanoate SMILES: CCC(=O)OCC(C)C