missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isobutyl Nitrite 95.0+%, TCI America™
Supplier: TCI America I0528100ML
Specifications
| Isobutyl Nitrite | |
| Yellow | |
| C4H9NO2 | |
| 100 mL | |
| isobutyl nitrite, isobutylnitrite, nitrous acid, 2-methylpropyl ester, nitrous acid, isobutyl ester, blackjack, isobutylnitrit, unii-gw9wab6qom, ccris 1099, ibn, nitrous acid 2-methylpropyl ester | |
| CC(C)CON=O | |
| 103.121 | |
| CHEBI:46643 | |
| ≥95.0% (GC) |
| 542-56-3 | |
| 67°C | |
| MFCD00002056 | |
| 2351 | |
| APNSGVMLAYLYCT-UHFFFAOYSA-N | |
| 2-methylpropyl nitrite | |
| 10958 | |
| 103.12 | |
| Liquid |
Chemical Identifiers
| 542-56-3 | |
| 103.121 | |
| APNSGVMLAYLYCT-UHFFFAOYSA-N | |
| 10958 | |
| 2-methylpropyl nitrite |
| C4H9NO2 | |
| MFCD00002056 | |
| isobutyl nitrite, isobutylnitrite, nitrous acid, 2-methylpropyl ester, nitrous acid, isobutyl ester, blackjack, isobutylnitrit, unii-gw9wab6qom, ccris 1099, ibn, nitrous acid 2-methylpropyl ester | |
| CHEBI:46643 | |
| CC(C)CON=O |
Safety and Handling
RTECSNumber : RA0805000
TSCA : Yes
Recommended Storage : Refrigerator