missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl Nitrite 95.0+%, TCI America™

Catalog No. p-7131461
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I052825ML 25 mL
I0528100ML 100 mL
I0528500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I052825ML Supplier TCI America Supplier No. I052825ML

Chemical Identifiers

CAS 542-56-3
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00002056
InChI Key APNSGVMLAYLYCT-UHFFFAOYSA-N
Synonym isobutyl nitrite, isobutylnitrite, nitrous acid, 2-methylpropyl ester, nitrous acid, isobutyl ester, blackjack, isobutylnitrit, unii-gw9wab6qom, ccris 1099, ibn, nitrous acid 2-methylpropyl ester
PubChem CID 10958
ChEBI CHEBI:46643
IUPAC Name 2-methylpropyl nitrite
SMILES CC(C)CON=O

Specifications

Color Yellow
Boiling Point 67°C
Quantity 25 mL
UN Number 2351
Formula Weight 103.12
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutyl Nitrite
RTECSNumber RA0805000
TSCA Yes
Recommended Storage Refrigerator