missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl n-Octanoate 99.0+%, TCI America™

Catalog No. O003225ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O003225ML 25 mL
O0032500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O003225ML Supplier TCI America Supplier No. O003225ML

Chemical Identifiers

CAS 5461-06-3
Molecular Formula C12H24O2
Molecular Weight (g/mol) 200.32
MDL Number MFCD00059444
InChI Key CFQRBRGFNFRMBD-UHFFFAOYSA-N
Synonym n-Octanoic Acid Isobutyl Ester
PubChem CID 79582
IUPAC Name 2-methylpropyl octanoate
SMILES CCCCCCCC(=O)OCC(C)C

Specifications

Color Yellow
Quantity 25 mL
Formula Weight 200.32
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutyl n-Octanoate
EINECSNumber (2)-0782
TSCA Yes