accessibility menu, dialog, popup

UserName

H3COOCH3CH3

CAS RN 5461-06-3

IUPAC Name: 2-methylpropyl octanoate
Synonyms: 2-methylpropyl octanoate
Molecular Weight (g/mol): 200.32
Molecular Formula: C12H24O2
InChi Key: CFQRBRGFNFRMBD-UHFFFAOYSA-N
SMILES: CCCCCCCC(=O)OCC(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results