missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl Myristate 97.0+%, TCI America™

Catalog No. M055525ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M055525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M055525ML Supplier TCI America Supplier No. M055525ML

Chemical Identifiers

CAS 25263-97-2
Molecular Formula C18H36O2
Molecular Weight (g/mol) 284.48
MDL Number MFCD00059286
InChI Key SKVCWXRLKHBEKW-UHFFFAOYSA-N
Synonym Myristic Acid Isobutyl Ester
PubChem CID 91361
IUPAC Name 2-methylpropyl tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)OCC(C)C

Specifications

Color Colorless
Quantity 25 mL
Formula Weight 284.48
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutyl Myristate
EINECSNumber (2)-0798
TSCA No